Learn More
ABclonal Technology Phospho-Histone H2AX-S139 Rabbit mAb 100 L
SDP
Supplier:  ABclonal Technology AP0687100UL
Encompass_Preferred
Immunogen: A synthetic phosphorylated peptide around S139 of human Histone H2A.X (P16104).•Host Species: Rabbit•Isotype: IgG•Species Reactivity:Human,Mouse,Rat•Validated Applications:WB,IHC•Swiss Prot: P16104•Gene ID:3014•Purification Method:Affinity purification•Storage: Store at -20℃. Avoid freeze / thaw cycles.|Buffer: PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.This product is available in 20UL, 50UL, 100UL, and 200UL. Please contact customer service for pricing and availability on alternative sizes.
Catalog No. NC2080004