Learn More
Cell Signaling Technology Phospho-Histone H3 (Ser10) Matched Antibody Pair, 1 Kit
SDP
Catalog No. 503132124
Encompass
The Phospho-Histone H3 (Ser10) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require custom antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, supporting applications such as fluorophore, lanthanide, biotin, or bead conjugation, and is compatible with platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Supplied in BSA and azide-free formulation
  • Compatible with diverse conjugation methods
  • Validated for reactivity in human, mouse, rat, and monkey samples
  • Optimized for high-throughput immunoassay platforms
  • Includes 100 μg each of capture and detection monoclonal antibodies
Catalog No. 50-313-2124 Supplier Cell Signaling Technology Supplier No. 59770S
The Phospho-Histone H3 (Ser10) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require custom antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, supporting applications such as fluorophore, lanthanide, biotin, or bead conjugation, and is compatible with platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Supplied in BSA and azide-free formulation
  • Compatible with diverse conjugation methods
  • Validated for reactivity in human, mouse, rat, and monkey samples
  • Optimized for high-throughput immunoassay platforms
  • Includes 100 μg each of capture and detection monoclonal antibodies