Learn More
Cell Signaling Technology Phospho-HS1 Matched Antibody Pair, Mouse, Unconjugated, 1 Kit
SDP
Catalog No. 503133881
Encompass
The Phospho-HS1 (Tyr378/397) Matched Antibody Pair is designed for use in immunoassays and high-throughput ELISA platforms, providing capture and detection antibodies that bind to non-overlapping epitopes. This kit is suitable for applications requiring custom antibody labeling, such as with fluorophores, lanthanides, biotin, or beads, and is compatible with platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex. The antibodies are provided in a BSA and azide-free formulation, making them ideal for specialized conjugation.
  • Validated for mouse species reactivity in ELISA applications
  • Consists of 100 μg each of capture and detection antibodies
  • BSA and azide-free formulation
  • Compatible with diverse immunoassay and high-throughput platforms
  • Optimal concentrations should be determined by the end user
Catalog No. 50-313-3881 Supplier Cell Signaling Technology Supplier No. 84046S
The Phospho-HS1 (Tyr378/397) Matched Antibody Pair is designed for use in immunoassays and high-throughput ELISA platforms, providing capture and detection antibodies that bind to non-overlapping epitopes. This kit is suitable for applications requiring custom antibody labeling, such as with fluorophores, lanthanides, biotin, or beads, and is compatible with platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex. The antibodies are provided in a BSA and azide-free formulation, making them ideal for specialized conjugation.
  • Validated for mouse species reactivity in ELISA applications
  • Consists of 100 μg each of capture and detection antibodies
  • BSA and azide-free formulation
  • Compatible with diverse immunoassay and high-throughput platforms
  • Optimal concentrations should be determined by the end user