Learn More
Cell Signaling Technology Matched Antibody Pair, Phospho-HS1 (Tyr378/397), Unconjugated, Human, 1 Kit
SDP
Catalog No. 503132362
Encompass
The Phospho-HS1 (Tyr378/397) Matched Antibody Pair II is designed for immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with specialized or custom labeling. This unconjugated kit provides essential tools for detecting phosphorylated HS1, a protein kinase substrate involved in actin dynamics and signaling complex formation following immune receptor stimulation.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl (pH 7.8))
  • BSA and azide free
  • Validated for human species reactivity
  • Consists of capture and detection antibodies that bind to non-overlapping epitopes
  • Compatible with diverse platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex
Catalog No. 50-313-2362 Supplier Cell Signaling Technology Supplier No. 89898S
The Phospho-HS1 (Tyr378/397) Matched Antibody Pair II is designed for immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with specialized or custom labeling. This unconjugated kit provides essential tools for detecting phosphorylated HS1, a protein kinase substrate involved in actin dynamics and signaling complex formation following immune receptor stimulation.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl (pH 7.8))
  • BSA and azide free
  • Validated for human species reactivity
  • Consists of capture and detection antibodies that bind to non-overlapping epitopes
  • Compatible with diverse platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex