Learn More
Enzo Life Sciences HSP 27 (pSer82) polyclonal antibody (50 µl)
SDP
Supplier:  Enzo Life Sciences ADISPA524D
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human HSP27 phosphorylated at Ser82. UniProt: P04792. Source: Whole rabbit serum. Species Reactivity: Human, Mouse, Rat, Sheep, Guinea pig, Dog, Chicken, Bovine. Applications: IF, IP, WB. Recommended dilutions: Immunoprecipitation (1:100). Western Blot (1:2,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Formulation: Liquid. Whole rabbit serum. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-468