Learn More
Cell Signaling Technology Phospho-IGF-I Receptor Beta Matched Antibody Pair, Human, 1 Kit
SDP
Catalog No. 503132956
Encompass
The Phospho-IGF-I Receptor β (Tyr1131) Matched Antibody Pair is designed for use in immunoassays and high-throughput platforms that require specialized or custom antibody labeling, including fluorophores, lanthanides, biotin, and beads. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, making it compatible with various platforms such as MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex.
  • Ideal for high-throughput ELISA platforms and immunoassay technologies
  • Compatible with labeling methods including fluorophores, lanthanides, biotin, and beads
  • Consists of capture and detection antibodies that bind to non-overlapping epitopes
  • Validated for human species reactivity
  • Supplied in 1X PBS (pH 7.8), BSA and azide free
Catalog No. 50-313-2956 Supplier Cell Signaling Technology Supplier No. 65853S
The Phospho-IGF-I Receptor β (Tyr1131) Matched Antibody Pair is designed for use in immunoassays and high-throughput platforms that require specialized or custom antibody labeling, including fluorophores, lanthanides, biotin, and beads. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, making it compatible with various platforms such as MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex.
  • Ideal for high-throughput ELISA platforms and immunoassay technologies
  • Compatible with labeling methods including fluorophores, lanthanides, biotin, and beads
  • Consists of capture and detection antibodies that bind to non-overlapping epitopes
  • Validated for human species reactivity
  • Supplied in 1X PBS (pH 7.8), BSA and azide free