Learn More
Cell Signaling Technology Phospho-IKK Alpha/Beta Antibody Sampler Kit, Phospho-Specific, Unconjugated, 1 Kit
SDP
Catalog No. 502729462
Encompass
The Phospho-IKK alpha/beta (Ser176/180) Antibody Sampler Kit provides a selection of antibodies to examine the levels of IKKalpha and IKKbeta proteins when phosphorylated at their key activation loop residues. This kit is designed for the detection of phosphorylation at Ser176/180 in IKKalpha and Ser177/181 in IKKbeta, which are critical regulatory events that trigger kinase activation within the NF-κB signaling pathway.
  • Includes primary antibodies for the analysis of phosphorylated IKKalpha and IKKbeta.
  • Contains an HRP-linked secondary antibody for convenient Western blot detection.
  • Optimized for the study of phosphorylation-dependent kinase activation and NF-κB signaling regulation.
  • Validated for multiple applications including Western blotting, immunohistochemistry, and flow cytometry.
  • Supplied in a stable formulation containing 50% glycerol.
Catalog No. 50-272-9462 Supplier Cell Signaling Technology Supplier No. 9958T
The Phospho-IKK alpha/beta (Ser176/180) Antibody Sampler Kit provides a selection of antibodies to examine the levels of IKKalpha and IKKbeta proteins when phosphorylated at their key activation loop residues. This kit is designed for the detection of phosphorylation at Ser176/180 in IKKalpha and Ser177/181 in IKKbeta, which are critical regulatory events that trigger kinase activation within the NF-κB signaling pathway.
  • Includes primary antibodies for the analysis of phosphorylated IKKalpha and IKKbeta.
  • Contains an HRP-linked secondary antibody for convenient Western blot detection.
  • Optimized for the study of phosphorylation-dependent kinase activation and NF-κB signaling regulation.
  • Validated for multiple applications including Western blotting, immunohistochemistry, and flow cytometry.
  • Supplied in a stable formulation containing 50% glycerol.