Learn More
Cell Signaling Technology Phospho-IKK Alpha Matched Antibody Pair, Human, 1 Kit
SDP
Catalog No. 503131865
Encompass
The Phospho-IKKα (Ser176/180) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with specialized or custom labeling. The kit includes capture and detection antibodies that bind to non-overlapping epitopes, facilitating flexible assay development.
  • Compatible with labeling methods including fluorophores, lanthanides, biotin, and beads
  • Suitable for platforms such as MSD, Quanterix Simoa, Alpha Technology, and Luminex
  • Includes one 100 μg vial each of capture and detection antibodies
  • BSA and azide-free formulation
  • Species reactivity confirmed for human
Catalog No. 50-313-1865 Supplier Cell Signaling Technology Supplier No. 67526S
The Phospho-IKKα (Ser176/180) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with specialized or custom labeling. The kit includes capture and detection antibodies that bind to non-overlapping epitopes, facilitating flexible assay development.
  • Compatible with labeling methods including fluorophores, lanthanides, biotin, and beads
  • Suitable for platforms such as MSD, Quanterix Simoa, Alpha Technology, and Luminex
  • Includes one 100 μg vial each of capture and detection antibodies
  • BSA and azide-free formulation
  • Species reactivity confirmed for human