Learn More
Cell Signaling Technology Phospho-Insulin Receptor Beta Matched Antibody Pair, Human, Unconjugated, 1 Kit
SDP
Catalog No. 503132047
Encompass
The Phospho-Insulin Receptor β (panTyr) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms. This kit provides capture and detection antibodies optimized for specialized or custom labeling with reagents such as fluorophores, lanthanides, biotin, or beads. It is suitable for platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Includes Insulin Receptor β (A6H3G) Rabbit Monoclonal Antibody and Phospho-Tyrosine (9E9) Mouse Monoclonal Antibody
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, 140 mM NaCl, pH 7.8)
  • BSA and azide free
  • Validated for human reactivity
  • Optimal for ELISA applications
  • Concentration: 1 mg/mL
Catalog No. 50-313-2047 Supplier Cell Signaling Technology Supplier No. 50239S
The Phospho-Insulin Receptor β (panTyr) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms. This kit provides capture and detection antibodies optimized for specialized or custom labeling with reagents such as fluorophores, lanthanides, biotin, or beads. It is suitable for platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Includes Insulin Receptor β (A6H3G) Rabbit Monoclonal Antibody and Phospho-Tyrosine (9E9) Mouse Monoclonal Antibody
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, 140 mM NaCl, pH 7.8)
  • BSA and azide free
  • Validated for human reactivity
  • Optimal for ELISA applications
  • Concentration: 1 mg/mL