Learn More
Enzo Life Sciences [pTyr1150/Tyr1151]Insulin receptor monoclonal antibody (10C3) (100 µg)
SDP
Supplier:  Enzo Life Sciences ADI905645100
Encompass_Preferred
Clone: 10C3. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to a portion of human Insulin Receptor phosphorylated at Tyr1150/1151, conjugated to KLH. UniProt: P06213. Species Reactivity: Human. Crossreactivity: Cross-reacts with IGF1 receptor. Applications: ELISA, WB. Recommended dilutions: ELISA (0.1µg/ml). Western Blot (0.2-0.5µg/ml, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Liquid. In PBS containing 50% glycerol, PEG, sucrose, and 0.09% sodium azide.
Catalog No. 50-658-401