Learn More
Cell Signaling Technology Phospho-IRF-3 (Ser379) Matched Antibody Pair, Mouse, Unconjugated, 1 Kit
SDP
Catalog No. 503131822
Encompass
The Phospho-IRF-3 (Ser379) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with specialized or custom labeling, such as fluorophores, lanthanides, biotin, or beads. This kit is suitable for applications requiring conjugated matched antibody pairs, including MSD, Quanterix Simoa, Alpha Technology, and Luminex, and specifically targets IRF-3 (Ser379) in mouse samples.
  • Designed for use with high-throughput ELISA platforms
  • Compatible with custom labeling (fluorophores, lanthanides, biotin, beads)
  • Supports research applications in platforms like MSD, Quanterix Simoa, and Alpha Technology
  • Validated for mouse species reactivity
  • Supplied in BSA and azide-free formulation
Catalog No. 50-313-1822 Supplier Cell Signaling Technology Supplier No. 67782S
The Phospho-IRF-3 (Ser379) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with specialized or custom labeling, such as fluorophores, lanthanides, biotin, or beads. This kit is suitable for applications requiring conjugated matched antibody pairs, including MSD, Quanterix Simoa, Alpha Technology, and Luminex, and specifically targets IRF-3 (Ser379) in mouse samples.
  • Designed for use with high-throughput ELISA platforms
  • Compatible with custom labeling (fluorophores, lanthanides, biotin, beads)
  • Supports research applications in platforms like MSD, Quanterix Simoa, and Alpha Technology
  • Validated for mouse species reactivity
  • Supplied in BSA and azide-free formulation