Learn More
Cell Signaling Technology Phospho-IRS-1 (Ser302) Matched Antibody Pair, Human Reactivity, 1 Kit
SDP
Catalog No. 503131936
Encompass
The Phospho-IRS-1 (Ser302) Matched Antibody Pair is designed for immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with specialized or custom labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes and are supplied in a BSA and azide-free formulation for maximum compatibility with downstream conjugation.
  • Validated for use in ELISA and high-throughput immunoassay platforms.
  • Compatible with a variety of labels including fluorophores, lanthanides, biotin, and beads.
  • Supplied as a complete pair containing both capture and detection antibodies.
  • Formulated in 1X PBS (pH 7.8) without BSA or sodium azide.
  • Species reactivity validated for human samples.
Catalog No. 50-313-1936 Supplier Cell Signaling Technology Supplier No. 26706S
The Phospho-IRS-1 (Ser302) Matched Antibody Pair is designed for immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with specialized or custom labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes and are supplied in a BSA and azide-free formulation for maximum compatibility with downstream conjugation.
  • Validated for use in ELISA and high-throughput immunoassay platforms.
  • Compatible with a variety of labels including fluorophores, lanthanides, biotin, and beads.
  • Supplied as a complete pair containing both capture and detection antibodies.
  • Formulated in 1X PBS (pH 7.8) without BSA or sodium azide.
  • Species reactivity validated for human samples.