Learn More
Cell Signaling Technology Phospho-Jak Family Antibody Sampler Kit, Unconjugated, 1 Kit
SDP
Catalog No. NC2754153
Encompass
The Phospho-Jak Family Antibody Sampler Kit offers a cost-effective method for monitoring the activation state of Janus family members through the use of both phospho-specific and total protein control antibodies. The kit is designed to provide sufficient antibody volumes for two western blot experiments per primary antibody target, facilitating the study of Jak-mediated signal transduction pathways.
  • Includes primary antibodies for Jak1, Jak2, Jak3, and Tyk2 in both total and phospho-specific formats.
  • Supplied with Anti-rabbit IgG, HRP-linked secondary antibody for convenient western blot detection.
  • Validated for multiple applications, including western blotting, immunoprecipitation, and immunohistochemistry (paraffin).
  • Formulated in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/ml BSA, 50% glycerol and less than 0.02% sodium azide.
Catalog No. NC2754153 Supplier Cell Signaling Technology Supplier No. 97999T
The Phospho-Jak Family Antibody Sampler Kit offers a cost-effective method for monitoring the activation state of Janus family members through the use of both phospho-specific and total protein control antibodies. The kit is designed to provide sufficient antibody volumes for two western blot experiments per primary antibody target, facilitating the study of Jak-mediated signal transduction pathways.
  • Includes primary antibodies for Jak1, Jak2, Jak3, and Tyk2 in both total and phospho-specific formats.
  • Supplied with Anti-rabbit IgG, HRP-linked secondary antibody for convenient western blot detection.
  • Validated for multiple applications, including western blotting, immunoprecipitation, and immunohistochemistry (paraffin).
  • Formulated in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/ml BSA, 50% glycerol and less than 0.02% sodium azide.