Learn More
Cell Signaling Technology Phospho-Jak2 Monoclonal Antibody, Rabbit, Human, Mouse, Rat, 100 μg
SDP
Catalog No. 503132904
Encompass
The Phospho-Jak2 (Tyr1007/1008) (C80C3) Rabbit Monoclonal Antibody is a high-quality antibody designed to detect endogenous levels of Jak2 when phosphorylated at Tyr1007 and Tyr1008. This carrier-free monoclonal antibody is produced by immunizing animals with a synthetic phosphopeptide and is formulated without BSA and azide, making it ideal for specialized applications such as fluorophore, metal, or oligonucleotide labeling.
  • Detects endogenous Jak2 phosphorylated at Tyr1007/1008
  • Suitable for Western blotting and ELISA
  • Validated for human, mouse, and rat reactivity
  • Carrier-free formulation optimized for custom labeling
  • Produced via immunization with a synthetic phosphopeptide
  • Maintains high integrity verified by size exclusion chromatography
Catalog No. 50-313-2904 Supplier Cell Signaling Technology Supplier No. 77546SF
The Phospho-Jak2 (Tyr1007/1008) (C80C3) Rabbit Monoclonal Antibody is a high-quality antibody designed to detect endogenous levels of Jak2 when phosphorylated at Tyr1007 and Tyr1008. This carrier-free monoclonal antibody is produced by immunizing animals with a synthetic phosphopeptide and is formulated without BSA and azide, making it ideal for specialized applications such as fluorophore, metal, or oligonucleotide labeling.
  • Detects endogenous Jak2 phosphorylated at Tyr1007/1008
  • Suitable for Western blotting and ELISA
  • Validated for human, mouse, and rat reactivity
  • Carrier-free formulation optimized for custom labeling
  • Produced via immunization with a synthetic phosphopeptide
  • Maintains high integrity verified by size exclusion chromatography