Learn More
Cell Signaling Technology Phospho-Jak2 Matched Antibody Pair, Human and Mouse Reactivity, 1 Kit
SDP
Catalog No. 503134226
Encompass
The Phospho-Jak2 (Tyr1007/1008) Matched Antibody Pair is designed for high-throughput ELISA platforms and immunoassay technologies that require custom or specialized antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes and are compatible with various conjugation methods including fluorophores, lanthanides, biotin, and beads.
  • Compatible with MSD, Quanterix Simoa, Alpha Technology, and Luminex platforms
  • Features antibodies ready for labeling with fluorophores, lanthanides, biotin, or beads
  • Supplied in 1X PBS (pH 7.8) with BSA and azide free
  • Validated for reactivity with human and mouse samples
  • Includes Jak2 (D5L7C) mouse monoclonal and phospho-Jak2 (Tyr1007/1008) (C80C3) rabbit monoclonal antibodies
Catalog No. 50-313-4226 Supplier Cell Signaling Technology Supplier No. 88866S
The Phospho-Jak2 (Tyr1007/1008) Matched Antibody Pair is designed for high-throughput ELISA platforms and immunoassay technologies that require custom or specialized antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes and are compatible with various conjugation methods including fluorophores, lanthanides, biotin, and beads.
  • Compatible with MSD, Quanterix Simoa, Alpha Technology, and Luminex platforms
  • Features antibodies ready for labeling with fluorophores, lanthanides, biotin, or beads
  • Supplied in 1X PBS (pH 7.8) with BSA and azide free
  • Validated for reactivity with human and mouse samples
  • Includes Jak2 (D5L7C) mouse monoclonal and phospho-Jak2 (Tyr1007/1008) (C80C3) rabbit monoclonal antibodies