Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-JNK1/2/3 (Thr183) Antibody | 10 UL
SDP

Catalog No. 5000442645
Encompass_Preferred

Phospho-JNK1/2/3 (Thr183) Antibody is a rabbit-derived and non-conjugated IgG polyclonal antibody, targeting Phospho-JNK1/2/3 (Thr183).

  • Host: Rabbit
  • Clonality: Polyclonal
  • Species reactivity: Human, mouse, rat
  • Immunogen: Synthetic phosphopeptide corresponding to residues surrounding Thr183 of human Phospho-JNK1/2/3
  • Purity: Affinity purified
  • Conjugation: Non-conjugated
  • Modification: Phosphorylated
  • Isotype: IgG
  • Appearance: Liquid
  • Formulation: Supplied in rabbit IgG in 10 mM phosphate buffered saline, pH 7.4, 150 mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol
  • Storage: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles
  • Shipping: Shipping with blue ice
  • Applications: WB, IHC-P, ICC/IF, IP, FC

Catalog No. 50-004-42645 Supplier Medchemexpress LLC Supplier No. HYP8083010UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phospho-JNK1/2/3 (Thr183) Antibody is a rabbit-derived and non-conjugated IgG polyclonal antibody, targeting Phospho-JNK1/2/3 (Thr183).

  • Host: Rabbit
  • Clonality: Polyclonal
  • Species reactivity: Human, mouse, rat
  • Immunogen: Synthetic phosphopeptide corresponding to residues surrounding Thr183 of human Phospho-JNK1/2/3
  • Purity: Affinity purified
  • Conjugation: Non-conjugated
  • Modification: Phosphorylated
  • Isotype: IgG
  • Appearance: Liquid
  • Formulation: Supplied in rabbit IgG in 10 mM phosphate buffered saline, pH 7.4, 150 mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol
  • Storage: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles
  • Shipping: Shipping with blue ice
  • Applications: WB, IHC-P, ICC/IF, IP, FC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.