Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-JNK1/2/3 (Thr183) Antibody | 100 UL
SDP

Catalog No. 5000441952
Encompass_Preferred

Phospho-JNK1/2/3 (Thr183) Antibody is a rabbit-derived and non-conjugated IgG polyclonal antibody, targeting to Phospho-JNK1/2/3 (Thr183). It is for research use only and not sold to patients.

  • Host: Rabbit
  • Clonality: Polyclonal
  • Species reactivity: Human, mouse, rat
  • Immunogen: Synthetic phosphopeptide (Thr183)
  • Applications: WB (1:500-1:1000), IHC-P (1:50-1:100), ICC/IF (1:50-1:200), IP (1:20), FC (1:50-1:100)
  • Sensitivity: Endogenous
  • Purity: Affinity purified
  • Conjugation: Non-conjugated
  • Modification: Phosphorylated
  • Isotype: IgG
  • Appearance: Liquid
  • Formulation: Rabbit IgG in PBS, 0.05% BSA, 0.02% sodium azide, 50% glycerol
  • Storage and stability: Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles

Catalog No. 50-004-41952 Supplier Medchemexpress LLC Supplier No. HYP80830100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phospho-JNK1/2/3 (Thr183) Antibody is a rabbit-derived and non-conjugated IgG polyclonal antibody, targeting to Phospho-JNK1/2/3 (Thr183). It is for research use only and not sold to patients.

  • Host: Rabbit
  • Clonality: Polyclonal
  • Species reactivity: Human, mouse, rat
  • Immunogen: Synthetic phosphopeptide (Thr183)
  • Applications: WB (1:500-1:1000), IHC-P (1:50-1:100), ICC/IF (1:50-1:200), IP (1:20), FC (1:50-1:100)
  • Sensitivity: Endogenous
  • Purity: Affinity purified
  • Conjugation: Non-conjugated
  • Modification: Phosphorylated
  • Isotype: IgG
  • Appearance: Liquid
  • Formulation: Rabbit IgG in PBS, 0.05% BSA, 0.02% sodium azide, 50% glycerol
  • Storage and stability: Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.