Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-JNK1/2/3 (Thr183) Antibody | 50 UL
SDP

Catalog No. 5000442195
Encompass_Preferred

Phospho-JNK1/2/3 (Thr183) Antibody is a rabbit-derived, non-conjugated IgG polyclonal antibody designed for research use. It targets Phospho-JNK1/2/3 (Thr183) and shows reactivity with human, mouse, and rat species.

  • Rabbit-derived, polyclonal IgG antibody
  • Targets Phospho-JNK1/2/3 (Thr183)
  • Reactivity with human, mouse, and rat
  • Predicted band size: 48 kDa; observed: 46/54 kDa
  • Affinity purified and non-conjugated
  • Suitable for WB, IHC-P, ICC/IF, IP, and FC applications
  • Ships with blue ice
  • Storage at -20°C for 1 year

Catalog No. 50-004-42195 Supplier Medchemexpress LLC Supplier No. HYP8083050UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phospho-JNK1/2/3 (Thr183) Antibody is a rabbit-derived, non-conjugated IgG polyclonal antibody designed for research use. It targets Phospho-JNK1/2/3 (Thr183) and shows reactivity with human, mouse, and rat species.

  • Rabbit-derived, polyclonal IgG antibody
  • Targets Phospho-JNK1/2/3 (Thr183)
  • Reactivity with human, mouse, and rat
  • Predicted band size: 48 kDa; observed: 46/54 kDa
  • Affinity purified and non-conjugated
  • Suitable for WB, IHC-P, ICC/IF, IP, and FC applications
  • Ships with blue ice
  • Storage at -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.