Learn More
Bioss Phospho-LRRK2 - Ser935 Recombi
SDP
Catalog No. 5000064903
Encompass_Preferred
LRRK2 Primary Antibody Unconjugated Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB IF(ICC) RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol
Catalog No. 50-000-64903 Supplier Bioss Supplier No. BSM62891R
LRRK2 Primary Antibody Unconjugated Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB IF(ICC) RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol