Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-MAPKAPK2 (Thr334) Antibody | 10 UL
SDP

Catalog No. 5000444170
Encompass_Preferred

The Phospho-MAPKAPK2 (Thr334) Antibody is a rabbit-derived antibody that reacts with human, mouse, and rat samples. It is supplied as a liquid at a concentration of 1.0 mg/mL and is non-conjugated. This antibody is suitable for applications such as Western Blot (WB), Immunohistochemistry-Paraffin (IHC-P), and Immunocytochemistry/Immunofluorescence (ICC/IF).

  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Concentration: 1.0 mg/mL
  • Appearance: Liquid
  • Conjugation: Non-conjugated
  • Formulation: Supplied in PBS containing 150 mM NaCl, 50% glycerol and 0.02% sodium azide, pH 7.4
  • Storage: Stored at -20°C for 1 year. Avoid repeated freeze / thaw cycles
  • Shipping: With blue ice
  • Applications: WB, IHC-P, ICC/IF
  • Dilution ratio: WB: 1:500-1:1000; IHC-P: 1:50-1:100; ICC/IF: 1:100-1:200
  • Predicted band size: 46 kDa
  • Observed band size: 49 kDa

Catalog No. 50-004-44170 Supplier Medchemexpress LLC Supplier No. HYP8377410UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Phospho-MAPKAPK2 (Thr334) Antibody is a rabbit-derived antibody that reacts with human, mouse, and rat samples. It is supplied as a liquid at a concentration of 1.0 mg/mL and is non-conjugated. This antibody is suitable for applications such as Western Blot (WB), Immunohistochemistry-Paraffin (IHC-P), and Immunocytochemistry/Immunofluorescence (ICC/IF).

  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Concentration: 1.0 mg/mL
  • Appearance: Liquid
  • Conjugation: Non-conjugated
  • Formulation: Supplied in PBS containing 150 mM NaCl, 50% glycerol and 0.02% sodium azide, pH 7.4
  • Storage: Stored at -20°C for 1 year. Avoid repeated freeze / thaw cycles
  • Shipping: With blue ice
  • Applications: WB, IHC-P, ICC/IF
  • Dilution ratio: WB: 1:500-1:1000; IHC-P: 1:50-1:100; ICC/IF: 1:100-1:200
  • Predicted band size: 46 kDa
  • Observed band size: 49 kDa

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.