Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-MAPKAPK2 (Thr334) Antibody | 100 UL
SDP

Catalog No. 5000443345
Encompass_Preferred

The Phospho-MAPKAPK2 (Thr334) Antibody is a non-conjugated rabbit antibody from MedChemExpress, designed to react with human, mouse, and rat samples. It is suitable for a variety of applications including Western Blot (WB), Immunohistochemistry (IHC-P), and Immunocytochemistry/Immunofluorescence (ICC/IF).

  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Concentration: 1.0 mg/mL
  • Applications: WB, IHC-P, ICC/IF
  • Predicted molecular weight: 46 kDa
  • Observed molecular weight: 49 kDa
  • Formulation: Supplied in PBS containing 150 mM NaCl, 50% glycerol and 0.02% sodium azide, pH 7.4
  • Storage: Store at -20°C for 1 year, avoid repeated freeze/thaw cycles

Catalog No. 50-004-43345 Supplier Medchemexpress LLC Supplier No. HYP83774100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Phospho-MAPKAPK2 (Thr334) Antibody is a non-conjugated rabbit antibody from MedChemExpress, designed to react with human, mouse, and rat samples. It is suitable for a variety of applications including Western Blot (WB), Immunohistochemistry (IHC-P), and Immunocytochemistry/Immunofluorescence (ICC/IF).

  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Concentration: 1.0 mg/mL
  • Applications: WB, IHC-P, ICC/IF
  • Predicted molecular weight: 46 kDa
  • Observed molecular weight: 49 kDa
  • Formulation: Supplied in PBS containing 150 mM NaCl, 50% glycerol and 0.02% sodium azide, pH 7.4
  • Storage: Store at -20°C for 1 year, avoid repeated freeze/thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.