Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-MAPKAPK2 (Thr334) Antibody | 50 UL
SDP

Catalog No. 5000444172
Encompass_Preferred

This liquid, non-conjugated antibody is suitable for WB, IHC-P, and ICC/IF applications. It is formulated in PBS containing 150 mM NaCl, 50% glycerol, and 0.02% sodium azide at pH 7.4.

  • Host: Rabbit
  • Reactivity: Human, Mouse, Rat
  • Recommended dilution ratios: WB: 1:500-1:1000; IHC-P: 1:50-1:100; ICC/IF: 1:100-1:200
  • Storage and stability: Stored at -20°C for 1 year, avoiding repeated freeze/thaw cycles
  • Shipping: Shipped with blue ice
  • SwissProt ID: P49137
  • Gene ID: 9261

Catalog No. 50-004-44172 Supplier Medchemexpress LLC Supplier No. HYP8377450UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This liquid, non-conjugated antibody is suitable for WB, IHC-P, and ICC/IF applications. It is formulated in PBS containing 150 mM NaCl, 50% glycerol, and 0.02% sodium azide at pH 7.4.

  • Host: Rabbit
  • Reactivity: Human, Mouse, Rat
  • Recommended dilution ratios: WB: 1:500-1:1000; IHC-P: 1:50-1:100; ICC/IF: 1:100-1:200
  • Storage and stability: Stored at -20°C for 1 year, avoiding repeated freeze/thaw cycles
  • Shipping: Shipped with blue ice
  • SwissProt ID: P49137
  • Gene ID: 9261

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.