Learn More
Cell Signaling Technology Phospho-Mer Matched Antibody Pair, Human, Unconjugated, 1 Kit
SDP
Catalog No. 503132331
Encompass
The Phospho-Mer (panTyr) Matched Antibody Pair is designed for use in high-throughput immunoassay and ELISA platforms that require specialized or custom antibody labeling, such as fluorophores, lanthanides, biotin, and beads. This pair of capture and detection antibodies is ideal for applications on platforms including MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex.
  • Supplied as a complete kit containing 100 μg each of rabbit monoclonal capture antibody and mouse monoclonal detection antibody
  • Optimized for versatility with conjugation-ready formats, including BSA and azide-free formulation
  • Validated for reactivity in human samples
  • Suitable for sensitive detection across various high-throughput screening technologies
Catalog No. 50-313-2331 Supplier Cell Signaling Technology Supplier No. 36532S
The Phospho-Mer (panTyr) Matched Antibody Pair is designed for use in high-throughput immunoassay and ELISA platforms that require specialized or custom antibody labeling, such as fluorophores, lanthanides, biotin, and beads. This pair of capture and detection antibodies is ideal for applications on platforms including MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex.
  • Supplied as a complete kit containing 100 μg each of rabbit monoclonal capture antibody and mouse monoclonal detection antibody
  • Optimized for versatility with conjugation-ready formats, including BSA and azide-free formulation
  • Validated for reactivity in human samples
  • Suitable for sensitive detection across various high-throughput screening technologies