Learn More
Medchemexpress LLC Phospho-Met c-Met antibody (YA175) | Protein A affinity purified | Predicted band size: 156 kDa; Observed band size: 170/140 kDa | 50 UL
SDP
Catalog No. 5000396034
Encompass_Preferred
MET is a receptor tyrosine kinase that transduces signals from the extracellular matrix into the cytoplasm by binding to hepatocyte growth factor (HGF) ligand. It regulates various physiological processes, including proliferation, scattering, morphogenesis, and survival. This antibody is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting Phospho-Met c-Met. It is supplied in 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol with 0.05% Sodium Azide as a preservative.
  • Targets Phospho-Met c-Met
  • Rabbit-derived monoclonal antibody
  • Reacts with human samples
  • Purified using protein A affinity
  • Applicable in Western blot (WB) and immunohistochemistry-paraffin (IHC-P)
  • For research use only
  • Avoid repeated freeze / thaw cycles
Catalog No. 50-003-96034 Supplier Medchemexpress LLC Supplier No. HYP8051250UL
MET is a receptor tyrosine kinase that transduces signals from the extracellular matrix into the cytoplasm by binding to hepatocyte growth factor (HGF) ligand. It regulates various physiological processes, including proliferation, scattering, morphogenesis, and survival. This antibody is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting Phospho-Met c-Met. It is supplied in 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol with 0.05% Sodium Azide as a preservative.
  • Targets Phospho-Met c-Met
  • Rabbit-derived monoclonal antibody
  • Reacts with human samples
  • Purified using protein A affinity
  • Applicable in Western blot (WB) and immunohistochemistry-paraffin (IHC-P)
  • For research use only
  • Avoid repeated freeze / thaw cycles