Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-Met c-Met (Tyr1349) antibody (YA213) | 100 UL
SDP

Catalog No. 5000442329
Encompass_Preferred

The Phospho-Met c-Met (Tyr1349) antibody (YA213) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting Phospho-Met c-Met (Tyr1349). This antibody is affinity purified and supplied in 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol, and 0.05% BSA with 0.01% Sodium azide as a preservative.

  • Recognizes Phospho-Met c-Met (Tyr1349)
  • Host species: Rabbit
  • Reactivity: Human
  • Research field: Signal transduction
  • Applications: Western blot (1:500-1:1000) and immunoprecipitation (1:20)
  • Molecular weight: Predicted 156 kDa, observed 170/140 kDa
  • Storage: Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles

Catalog No. 50-004-42329 Supplier Medchemexpress LLC Supplier No. HYP80803100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Phospho-Met c-Met (Tyr1349) antibody (YA213) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting Phospho-Met c-Met (Tyr1349). This antibody is affinity purified and supplied in 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol, and 0.05% BSA with 0.01% Sodium azide as a preservative.

  • Recognizes Phospho-Met c-Met (Tyr1349)
  • Host species: Rabbit
  • Reactivity: Human
  • Research field: Signal transduction
  • Applications: Western blot (1:500-1:1000) and immunoprecipitation (1:20)
  • Molecular weight: Predicted 156 kDa, observed 170/140 kDa
  • Storage: Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.