Learn More
Cell Signaling Technology Phospho-Met (panTyr) Matched Antibody Pair, Human, Unconjugated, 1 Kit
SDP
Catalog No. 503132256
Encompass
The Phospho-Met (panTyr) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs suitable for custom labeling, including conjugation with fluorophores, lanthanides, biotin, or beads. Compatible with platforms such as MSD, Quanterix Simoa, Luminex, and Alpha Technology, this kit provides capture and detection antibodies that bind to non-overlapping epitopes.
  • Supplied in BSA and azide-free formulation
  • Includes 100 μg each of Met (D1C2) Rabbit Monoclonal Antibody and Phospho-Tyrosine (9E9) Mouse Monoclonal Antibody
  • Validated for ELISA applications
  • Species reactivity: human
  • Requires storage at -20°C
Catalog No. 50-313-2256 Supplier Cell Signaling Technology Supplier No. 52467S
The Phospho-Met (panTyr) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs suitable for custom labeling, including conjugation with fluorophores, lanthanides, biotin, or beads. Compatible with platforms such as MSD, Quanterix Simoa, Luminex, and Alpha Technology, this kit provides capture and detection antibodies that bind to non-overlapping epitopes.
  • Supplied in BSA and azide-free formulation
  • Includes 100 μg each of Met (D1C2) Rabbit Monoclonal Antibody and Phospho-Tyrosine (9E9) Mouse Monoclonal Antibody
  • Validated for ELISA applications
  • Species reactivity: human
  • Requires storage at -20°C