Learn More
Cell Signaling Technology Phospho-MLKL (Ser358) Matched Antibody Pair, Human, Unconjugated, 1 Kit
SDP
Catalog No. 503131567
Encompass
The Phospho-MLKL (Ser358) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require custom antibody labeling, including conjugation to fluorophores, lanthanides, beads, or biotin. This pair is compatible with various detection systems, including MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex.
  • Includes capture and detection antibodies that bind to non-overlapping epitopes
  • Supplied in 1X PBS (pH 7.8)
  • BSA and azide free formulation
  • Validated for use in ELISA
  • Species reactivity: Human
Catalog No. 50-313-1567 Supplier Cell Signaling Technology Supplier No. 97985S
The Phospho-MLKL (Ser358) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require custom antibody labeling, including conjugation to fluorophores, lanthanides, beads, or biotin. This pair is compatible with various detection systems, including MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex.
  • Includes capture and detection antibodies that bind to non-overlapping epitopes
  • Supplied in 1X PBS (pH 7.8)
  • BSA and azide free formulation
  • Validated for use in ELISA
  • Species reactivity: Human