Learn More
Cell Signaling Technology Phospho-mTOR (Ser2481) Matched Antibody Pair, Human and Mouse, 1 Kit
SDP
Catalog No. 503131779
Encompass
The Phospho-mTOR (Ser2481) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling. This kit facilitates sensitive detection of mTOR phosphorylated at Ser2481, which is a key kinase involved in cell growth and homeostasis. The pair includes capture and detection antibodies formatted for compatibility with various labeling methods, including fluorophores, lanthanides, biotin, and beads.
  • Includes capture and detection antibodies for mTOR (Ser2481) detection
  • Compatible with platforms such as MSD, Quanterix Simoa, Alpha Technology, and Luminex
  • Supplied in BSA and azide-free formulation
  • Validated for human and mouse species reactivity
  • Ideal for applications requiring custom labeling
Catalog No. 50-313-1779 Supplier Cell Signaling Technology Supplier No. 16730S
The Phospho-mTOR (Ser2481) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling. This kit facilitates sensitive detection of mTOR phosphorylated at Ser2481, which is a key kinase involved in cell growth and homeostasis. The pair includes capture and detection antibodies formatted for compatibility with various labeling methods, including fluorophores, lanthanides, biotin, and beads.
  • Includes capture and detection antibodies for mTOR (Ser2481) detection
  • Compatible with platforms such as MSD, Quanterix Simoa, Alpha Technology, and Luminex
  • Supplied in BSA and azide-free formulation
  • Validated for human and mouse species reactivity
  • Ideal for applications requiring custom labeling