Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-NF-KB p105/p50(Ser337) antibody | 100 UL
SDP

Catalog No. 5000442684
Encompass_Preferred

Phospho-NF-KB p105/p50(Ser337) antibody is a Rabbit-derived and non-conjugated IgG polyclonal antibody, specifically targeting Phospho-NF-KB p105/p50(Ser337). It is intended for research use only.

  • Host: Rabbit
  • Clonality: Polyclonal
  • Conjugation: Non-conjugated
  • Modification: Phosphorylated
  • Isotype: IgG
  • Appearance: Liquid
  • Formulation: Supplied in 0.01M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% Glycerol
  • Storage & stability: Stored at -20°C for 1 year
  • Immunogen: KLH conjugated Synthesised phosphopeptide derived from human NF KappaB p105 around the phosphorylation site of Ser337: RK(p-S)DL
  • Species reactivity: Human, Mouse. Predicted reactivity: Rat, Pig, Cow
  • Sensitivity: Endogenous

Catalog No. 50-004-42684 Supplier Medchemexpress LLC Supplier No. HYP81049100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phospho-NF-KB p105/p50(Ser337) antibody is a Rabbit-derived and non-conjugated IgG polyclonal antibody, specifically targeting Phospho-NF-KB p105/p50(Ser337). It is intended for research use only.

  • Host: Rabbit
  • Clonality: Polyclonal
  • Conjugation: Non-conjugated
  • Modification: Phosphorylated
  • Isotype: IgG
  • Appearance: Liquid
  • Formulation: Supplied in 0.01M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% Glycerol
  • Storage & stability: Stored at -20°C for 1 year
  • Immunogen: KLH conjugated Synthesised phosphopeptide derived from human NF KappaB p105 around the phosphorylation site of Ser337: RK(p-S)DL
  • Species reactivity: Human, Mouse. Predicted reactivity: Rat, Pig, Cow
  • Sensitivity: Endogenous

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.