Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-NF-KB p105/p50(Ser337) Antibody | 50 UL
SDP

Catalog No. 5000442585
Encompass_Preferred

Phospho-NF-KB p105/p50(Ser337) Antibody is a rabbit-derived, non-conjugated IgG polyclonal antibody specifically targeting Phospho-NF-KB p105/p50(Ser337). This affinity-purified antibody is intended for research use only. NF-kappa-B is a crucial transcription factor involved in inflammation, immunity, cell growth, and other essential biological processes across various cell types.

  • Host: Rabbit
  • Clonality: Polyclonal
  • Reactivity: Human, mouse (predicted: rat, pig, cow)
  • Applications: WB, IHC-P, IHC-F, FC, ICC/IF, ELISA
  • Sensitivity: Endogenous
  • Conjugation: Non-conjugated
  • Modification: Phosphorylated
  • Isotype: IgG
  • Formulation: 0.01M TBS (pH7.4) with 1% BSA, 0.02% Proclin300, 50% Glycerol
  • Storage: Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Shipping: With blue ice
  • Purity: Affinity purified

Catalog No. 50-004-42585 Supplier Medchemexpress LLC Supplier No. HYP8104950UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phospho-NF-KB p105/p50(Ser337) Antibody is a rabbit-derived, non-conjugated IgG polyclonal antibody specifically targeting Phospho-NF-KB p105/p50(Ser337). This affinity-purified antibody is intended for research use only. NF-kappa-B is a crucial transcription factor involved in inflammation, immunity, cell growth, and other essential biological processes across various cell types.

  • Host: Rabbit
  • Clonality: Polyclonal
  • Reactivity: Human, mouse (predicted: rat, pig, cow)
  • Applications: WB, IHC-P, IHC-F, FC, ICC/IF, ELISA
  • Sensitivity: Endogenous
  • Conjugation: Non-conjugated
  • Modification: Phosphorylated
  • Isotype: IgG
  • Formulation: 0.01M TBS (pH7.4) with 1% BSA, 0.02% Proclin300, 50% Glycerol
  • Storage: Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Shipping: With blue ice
  • Purity: Affinity purified

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.