Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-NF-KB p65 (Ser536) Antibody | 10 UL
SDP

Catalog No. 5000442499
Encompass_Preferred

Phospho-NF-KB p65 (Ser536) Antibody is a Rabbit-derived, non-conjugated IgG polyclonal antibody specifically targeting Phospho-NF-KB p65 (Ser536). This antibody is for research use only and is supplied in 1*PBS (pH 7.3), 50% glycerol, and 0.5% BSA, with 0.02% sodium azide as a preservative.

  • Host: Rabbit
  • Clonality: Polyclonal
  • Isotype: IgG
  • Modification: Phosphorylated
  • Purity: Affinity purified
  • Predicted molecular weight: 60 kDa
  • Observed molecular weight: 65 kDa
  • Species reactivity: Human, mouse, rat
  • Applications: Western blot (1:500-1:1000), immunohistochemistry-paraffin (1:50-1:100), immunocytochemistry/immunofluorescence (1:50-1:200)

Catalog No. 50-004-42499 Supplier Medchemexpress LLC Supplier No. HYP8083910UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phospho-NF-KB p65 (Ser536) Antibody is a Rabbit-derived, non-conjugated IgG polyclonal antibody specifically targeting Phospho-NF-KB p65 (Ser536). This antibody is for research use only and is supplied in 1*PBS (pH 7.3), 50% glycerol, and 0.5% BSA, with 0.02% sodium azide as a preservative.

  • Host: Rabbit
  • Clonality: Polyclonal
  • Isotype: IgG
  • Modification: Phosphorylated
  • Purity: Affinity purified
  • Predicted molecular weight: 60 kDa
  • Observed molecular weight: 65 kDa
  • Species reactivity: Human, mouse, rat
  • Applications: Western blot (1:500-1:1000), immunohistochemistry-paraffin (1:50-1:100), immunocytochemistry/immunofluorescence (1:50-1:200)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.