Learn More
Medchemexpress LLC Phospho-NF-KB p65 (Ser536) Antibody | 50 UL
SDP
Catalog No. 5000442095
Encompass_Preferred
This antibody is a rabbit-derived, non-conjugated IgG polyclonal antibody designed to target Phospho-NF-KB p65 (Ser536). It is suitable for a variety of applications including Western Blot (WB), Immunohistochemistry-Paraffin (IHC-P), and Immunocytochemistry/Immunofluorescence (ICC/IF), reacting with human, mouse, and rat samples.
  • Rabbit-derived host
  • Non-conjugated IgG polyclonal antibody
  • Verified for Western Blot, IHC-P, and ICC/IF applications
  • Concentration of 1 mg/mL
  • Formulated in 1*PBS (pH 7.3), 50% glycerol and 0.5% BSA with 0.02% sodium azide
  • Stable for 1 year when stored at -20°C
  • Avoids repeated freeze/thaw cycles to maintain stability
  • Shipped with blue ice
Catalog No. 50-004-42095 Supplier Medchemexpress LLC Supplier No. HYP8083950UL
This antibody is a rabbit-derived, non-conjugated IgG polyclonal antibody designed to target Phospho-NF-KB p65 (Ser536). It is suitable for a variety of applications including Western Blot (WB), Immunohistochemistry-Paraffin (IHC-P), and Immunocytochemistry/Immunofluorescence (ICC/IF), reacting with human, mouse, and rat samples.
  • Rabbit-derived host
  • Non-conjugated IgG polyclonal antibody
  • Verified for Western Blot, IHC-P, and ICC/IF applications
  • Concentration of 1 mg/mL
  • Formulated in 1*PBS (pH 7.3), 50% glycerol and 0.5% BSA with 0.02% sodium azide
  • Stable for 1 year when stored at -20°C
  • Avoids repeated freeze/thaw cycles to maintain stability
  • Shipped with blue ice