Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-NF-KB p65 (Thr254) Antibody | 50 UL
SDP

Catalog No. 5000442647
Encompass_Preferred

The Phospho-NF-KB p65 (Thr254) Antibody is a rabbit-derived, non-conjugated IgG polyclonal antibody designed to target Phospho-NF-KB p65 (Thr254). It is supplied as a liquid in 1xPBS (pH 7.3), 50% glycerol, and 0.5% BSA, with 0.02% sodium azide as a preservative, making it suitable for various research applications.

  • Targets Phospho-NF-KB p65 (Thr254)
  • Rabbit-derived IgG polyclonal antibody
  • Non-conjugated formulation
  • Supplied in 1xPBS (pH 7.3), 50% glycerol, 0.5% BSA
  • Preservative: 0.02% sodium azide
  • Suitable for research use only
  • Applications: WB (1:500-1:1000), IHC-P (1:50-1:100), ICC/IF (1:50-1:200)
  • Species reactivity: Human, Mouse, Rat
  • Storage at -20°C for 1 year, avoid repeated freeze/thaw cycles

Catalog No. 50-004-42647 Supplier Medchemexpress LLC Supplier No. HYP8084050UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Phospho-NF-KB p65 (Thr254) Antibody is a rabbit-derived, non-conjugated IgG polyclonal antibody designed to target Phospho-NF-KB p65 (Thr254). It is supplied as a liquid in 1xPBS (pH 7.3), 50% glycerol, and 0.5% BSA, with 0.02% sodium azide as a preservative, making it suitable for various research applications.

  • Targets Phospho-NF-KB p65 (Thr254)
  • Rabbit-derived IgG polyclonal antibody
  • Non-conjugated formulation
  • Supplied in 1xPBS (pH 7.3), 50% glycerol, 0.5% BSA
  • Preservative: 0.02% sodium azide
  • Suitable for research use only
  • Applications: WB (1:500-1:1000), IHC-P (1:50-1:100), ICC/IF (1:50-1:200)
  • Species reactivity: Human, Mouse, Rat
  • Storage at -20°C for 1 year, avoid repeated freeze/thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.