Learn More
ABclonal Technology Phospho-NFKB1-S337 Rabbit
SDP
Catalog No. 5000013832
Encompass_Preferred
This gene encodes a 105 kD protein that is processed by the 26S proteasome into a 50 kD protein The 105 kD protein inhibits Rel protein-specific transcription while the 50 kD protein is a DNA-binding subunit of the NF-kappa-B (NFKB) complex NFKB is activated by various stimuli like cytokines free radicals UV irradiation and pathogens Once activated NFKB translocates to the nucleus to regulate genes involved in many biological functions Dysregulation of NFKB activation is linked to inflammatory diseases immune cell development issues and impaired cell growth NFKB is key in the immediate-early response to viral infections Alternative splicing produces multiple isoforms including one that undergoes proteolytic processing
Catalog No. 50-000-13832 Supplier ABclonal Technology Supplier No. AP0125PM20UL
This gene encodes a 105 kD protein that is processed by the 26S proteasome into a 50 kD protein The 105 kD protein inhibits Rel protein-specific transcription while the 50 kD protein is a DNA-binding subunit of the NF-kappa-B (NFKB) complex NFKB is activated by various stimuli like cytokines free radicals UV irradiation and pathogens Once activated NFKB translocates to the nucleus to regulate genes involved in many biological functions Dysregulation of NFKB activation is linked to inflammatory diseases immune cell development issues and impaired cell growth NFKB is key in the immediate-early response to viral infections Alternative splicing produces multiple isoforms including one that undergoes proteolytic processing