Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Phospho-NMDA Receptor 2A Monoclonal Antibody, Rabbit, 100 μl
SDP

Catalog No. 503131525
Encompass

The Phospho-NMDA Receptor 2A (GluN2A) (Tyr1246) (E4U3T) Rabbit Monoclonal Antibody is a recombinant antibody engineered to detect endogenous levels of the NMDA Receptor 2A protein, specifically when phosphorylated at the Tyr1246 residue. Developed by immunizing animals with a synthetic phosphopeptide, this monoclonal antibody may also cross-react with NMDA Receptor 2B (GluN2B) when phosphorylated at the conserved Tyr1252 site. It is designed for applications such as Western blotting to study receptor signaling pathways.

  • Validated for endogenous protein detection
  • Specific for phosphorylated Tyr1246 in GluN2A
  • Reactivity with phosphorylated Tyr1252 in GluN2B
  • Optimized for Western blotting applications
  • Host species: rabbit

Catalog No. 50-313-1525 Supplier Cell Signaling Technology Supplier No. 82049S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Phospho-NMDA Receptor 2A (GluN2A) (Tyr1246) (E4U3T) Rabbit Monoclonal Antibody is a recombinant antibody engineered to detect endogenous levels of the NMDA Receptor 2A protein, specifically when phosphorylated at the Tyr1246 residue. Developed by immunizing animals with a synthetic phosphopeptide, this monoclonal antibody may also cross-react with NMDA Receptor 2B (GluN2B) when phosphorylated at the conserved Tyr1252 site. It is designed for applications such as Western blotting to study receptor signaling pathways.

  • Validated for endogenous protein detection
  • Specific for phosphorylated Tyr1246 in GluN2A
  • Reactivity with phosphorylated Tyr1252 in GluN2B
  • Optimized for Western blotting applications
  • Host species: rabbit

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.