Learn More
ABclonal Technology Phospho-p38 MAPK-T180/Y182
SDP
Catalog No. 5000013946
Encompass_Preferred
This gene encodes a member of a family of protein kinases that are involved in the integration of biochemical signals for a wide variety of cellular processes including cell proliferation differentiation transcriptional regulation and development The encoded protein can be activated by proinflammatory cytokines and environmental stresses through phosphorylation by mitogen activated protein kinase kinases (MKKs) Alternative splicing results in multiple transcript variants [provided by RefSeq Mar 2014]
Catalog No. 50-000-13946 Supplier ABclonal Technology Supplier No. AP131120UL
This gene encodes a member of a family of protein kinases that are involved in the integration of biochemical signals for a wide variety of cellular processes including cell proliferation differentiation transcriptional regulation and development The encoded protein can be activated by proinflammatory cytokines and environmental stresses through phosphorylation by mitogen activated protein kinase kinases (MKKs) Alternative splicing results in multiple transcript variants [provided by RefSeq Mar 2014]