Learn More
Meso Scale Discovery PHOSPHO-P38 WHOLE CELL LYSATE
SDP
Catalog No. NC1838888
Encompass
Phospho-p38 Whole Cell Lysate Kit5 Plates. Assay-specific components for the quantitative determination of phospho-p38 (Thr180, Tyr182) in human, mouse, and rat whole cell lysates.
Catalog No. NC1838888 Supplier Meso Scale Discovery Supplier No. K150CYD2
Phospho-p38 Whole Cell Lysate Kit5 Plates. Assay-specific components for the quantitative determination of phospho-p38 (Thr180, Tyr182) in human, mouse, and rat whole cell lysates.