Learn More
Cell Signaling Technology Matched Antibody Pair, Phospho-p53 Ser15, Human, 1 Kit
SDP
Catalog No. 503133759
Encompass
The Phospho-p53 (Ser15) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling, including conjugation with fluorophores, lanthanides, biotin, or beads. This antibody pair is compatible with platforms such as MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex.
  • Ideal for high-throughput ELISA and immunoassay development.
  • Compatible with custom labeling methods including fluorophores and beads.
  • Supplied in 1X PBS, BSA and azide free for maximum versatility in downstream applications.
  • Validated for human species reactivity.
  • Capture and detection antibodies bind to non-overlapping epitopes.
Catalog No. 50-313-3759 Supplier Cell Signaling Technology Supplier No. 88007S
The Phospho-p53 (Ser15) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling, including conjugation with fluorophores, lanthanides, biotin, or beads. This antibody pair is compatible with platforms such as MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex.
  • Ideal for high-throughput ELISA and immunoassay development.
  • Compatible with custom labeling methods including fluorophores and beads.
  • Supplied in 1X PBS, BSA and azide free for maximum versatility in downstream applications.
  • Validated for human species reactivity.
  • Capture and detection antibodies bind to non-overlapping epitopes.