Learn More
BIOHIPPO Phospho-p53 Ser15 rAb W-R58
SDP
Catalog No. 5000975495
Encompass
U-Blot Phospho-p53 (Ser15) Mouse mAb is a Mouse-derived recombinant monoclonal antibody (clone W-R58) that specifically recognizes Phospho-p53 (Ser15) a phosphorylation modification at Ser15 It shows reactivity with Human Validated applications include WB ICC/IF Recommended dilutions WB 1500-11000 ICC/IF 150 Purified by protein a Tumor protein P53 also known as p53 cellular tumor antigen p53 is an isoform of a protein encoded by homologous genes in various organisms p53 is crucial in multicellular vertebrates where it prevents cancer formation As such p53 has been described as the guardian of the genome because of its role in conserving stability by preventing genome mutation Hence TP53 is classified as a tumor suppressor gene Supplied as 100 uL
Catalog No. 50-009-75495 Supplier BIOHIPPO Supplier No. BHA21821370
U-Blot Phospho-p53 (Ser15) Mouse mAb is a Mouse-derived recombinant monoclonal antibody (clone W-R58) that specifically recognizes Phospho-p53 (Ser15) a phosphorylation modification at Ser15 It shows reactivity with Human Validated applications include WB ICC/IF Recommended dilutions WB 1500-11000 ICC/IF 150 Purified by protein a Tumor protein P53 also known as p53 cellular tumor antigen p53 is an isoform of a protein encoded by homologous genes in various organisms p53 is crucial in multicellular vertebrates where it prevents cancer formation As such p53 has been described as the guardian of the genome because of its role in conserving stability by preventing genome mutation Hence TP53 is classified as a tumor suppressor gene Supplied as 100 uL