Learn More
Medchemexpress LLC Phospho-p53 (Ser6) Antibody (YA166) | 10 UL
SDP
Catalog No. 5000442500
Encompass_Preferred
Phospho-p53 (Ser6) Antibody (YA166) is a rabbit-derived, non-conjugated IgG monoclonal antibody designed to target Phospho-p53 (Ser6). This affinity-purified antibody is suitable for various research applications including Western Blot (WB), Immunohistochemistry (IHC-P), and Immunoprecipitation (IP).
  • Rabbit-derived monoclonal antibody
  • Targets Phospho-p53 (Ser6)
  • Non-conjugated IgG
  • Reacts with human samples
  • Suitable for Western Blot, Immunohistochemistry, and Immunoprecipitation
  • Store at -20°C for up to 1 year; avoid repeated freeze/thaw cycles
Catalog No. 50-004-42500 Supplier Medchemexpress LLC Supplier No. HYP8084410UL
Phospho-p53 (Ser6) Antibody (YA166) is a rabbit-derived, non-conjugated IgG monoclonal antibody designed to target Phospho-p53 (Ser6). This affinity-purified antibody is suitable for various research applications including Western Blot (WB), Immunohistochemistry (IHC-P), and Immunoprecipitation (IP).
  • Rabbit-derived monoclonal antibody
  • Targets Phospho-p53 (Ser6)
  • Non-conjugated IgG
  • Reacts with human samples
  • Suitable for Western Blot, Immunohistochemistry, and Immunoprecipitation
  • Store at -20°C for up to 1 year; avoid repeated freeze/thaw cycles