Learn More
Cell Signaling Technology Phospho-PDGF Receptor Alpha/Beta Rabbit Monoclonal Antibody, 100 μg
SDP
Catalog No. 503132653
Encompass
The Phospho-PDGF Receptor alpha (Tyr849)/PDGF Receptor beta (Tyr857) (C43E9) Rabbit Monoclonal Antibody is a highly specific, recombinant antibody designed to detect endogenous levels of PDGF receptor α and β only when phosphorylated at Tyr849 and Tyr857, respectively. This BSA and azide-free formulation is ideal for labeling applications including fluorophores, metals, lanthanides, and oligonucleotides.
  • Validated for western blotting applications.
  • Detects endogenous phosphorylation in human, mouse, and rat samples.
  • Carrier-free formulation optimized for custom labeling.
  • Quality control tested by size exclusion chromatography for antibody integrity.
Catalog No. 50-313-2653 Supplier Cell Signaling Technology Supplier No. 68236SF
The Phospho-PDGF Receptor alpha (Tyr849)/PDGF Receptor beta (Tyr857) (C43E9) Rabbit Monoclonal Antibody is a highly specific, recombinant antibody designed to detect endogenous levels of PDGF receptor α and β only when phosphorylated at Tyr849 and Tyr857, respectively. This BSA and azide-free formulation is ideal for labeling applications including fluorophores, metals, lanthanides, and oligonucleotides.
  • Validated for western blotting applications.
  • Detects endogenous phosphorylation in human, mouse, and rat samples.
  • Carrier-free formulation optimized for custom labeling.
  • Quality control tested by size exclusion chromatography for antibody integrity.