Learn More
Bioss Phospho-PDGF Receptor beta
SDP
Catalog No. 5000066176
Encompass_Preferred
PDGF Receptor beta Primary Antibody Unconjugated Host Species Rabbit Target Species Mouse Clonality Monoclonal Applications WB RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol
Catalog No. 50-000-66176 Supplier Bioss Supplier No. BSM62855R
PDGF Receptor beta Primary Antibody Unconjugated Host Species Rabbit Target Species Mouse Clonality Monoclonal Applications WB RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol