Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-PDGFR beta (Tyr740) Antibody (YA3062) | 10 UL
SDP

Catalog No. 5000443212
Encompass_Preferred

Phospho-PDGFR beta (Tyr740) Antibody (YA3062) is a rabbit-derived, non-conjugated IgG monoclonal antibody that targets Phospho-PDGFR beta (Tyr740). PDGF Receptor beta is a tyrosine-protein kinase functioning as a cell-surface receptor for homodimeric PDGFB and PDGFD, and heterodimers formed by PDGFA and PDGFB. It plays essential roles in embryonic development, cell proliferation, survival, differentiation, chemotaxis, and migration, with critical involvement in blood vessel development and cardiovascular system formation.

  • Rabbit-derived host
  • Reactivity: Mouse, Rat
  • Non-conjugated IgG monoclonal antibody
  • Applications: WB, ICC/IF
  • Purity by affinity chromatography
  • Supplied in 10 mM PBS, pH 7.4, 150 mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol
  • Stored at -20°C for 1 year; avoid repeated freeze/thaw cycles
  • Liquid appearance, shipped with blue ice
  • Concentration: 0.6 mg/mL

Catalog No. 50-004-43212 Supplier Medchemexpress LLC Supplier No. HYP8331710UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phospho-PDGFR beta (Tyr740) Antibody (YA3062) is a rabbit-derived, non-conjugated IgG monoclonal antibody that targets Phospho-PDGFR beta (Tyr740). PDGF Receptor beta is a tyrosine-protein kinase functioning as a cell-surface receptor for homodimeric PDGFB and PDGFD, and heterodimers formed by PDGFA and PDGFB. It plays essential roles in embryonic development, cell proliferation, survival, differentiation, chemotaxis, and migration, with critical involvement in blood vessel development and cardiovascular system formation.

  • Rabbit-derived host
  • Reactivity: Mouse, Rat
  • Non-conjugated IgG monoclonal antibody
  • Applications: WB, ICC/IF
  • Purity by affinity chromatography
  • Supplied in 10 mM PBS, pH 7.4, 150 mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol
  • Stored at -20°C for 1 year; avoid repeated freeze/thaw cycles
  • Liquid appearance, shipped with blue ice
  • Concentration: 0.6 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.