Learn More
Medchemexpress LLC Phospho-PDGFR beta (Y740) antibody | Protein A affinity purified. | 124 kDa | 10 UL
SDP
Catalog No. 5000395717
Encompass_Preferred
Phospho-PDGFR beta (Y740) Antibody (YA164) is a highly specific rabbit-derived IgG monoclonal antibody, targeting Phospho-PDGFR beta (Y740). This antibody is suitable for Western Blot (WB) and Immunocytochemistry/Immunofluorescence (ICC/IF) applications, offering reliable detection in human and rat samples. It is supplied in a liquid formulation containing TBS, BSA, and glycerol, ensuring stability and performance for various research needs.
  • Rabbit-derived IgG monoclonal antibody
  • Targets Phospho-PDGFR beta (Y740)
  • Suitable for WB and ICC/IF applications
  • Reactive with human and rat samples
  • Protein A affinity purified
  • Predicted band size of 124 kDa
  • Shipped with blue ice and stored at -20°C
Catalog No. 50-003-95717 Supplier Medchemexpress LLC Supplier No. HYP8027810UL
Phospho-PDGFR beta (Y740) Antibody (YA164) is a highly specific rabbit-derived IgG monoclonal antibody, targeting Phospho-PDGFR beta (Y740). This antibody is suitable for Western Blot (WB) and Immunocytochemistry/Immunofluorescence (ICC/IF) applications, offering reliable detection in human and rat samples. It is supplied in a liquid formulation containing TBS, BSA, and glycerol, ensuring stability and performance for various research needs.
  • Rabbit-derived IgG monoclonal antibody
  • Targets Phospho-PDGFR beta (Y740)
  • Suitable for WB and ICC/IF applications
  • Reactive with human and rat samples
  • Protein A affinity purified
  • Predicted band size of 124 kDa
  • Shipped with blue ice and stored at -20°C