Learn More
ABclonal Technology Phospho-PDK1/PDPK1-S241
SDP
Supplier:  ABclonal Technology AP1357100UL
Encompass_Preferred
Enables 3-phosphoinositide-dependent protein kinase activity phospholipase activator activity and phospholipase binding activity Involved in several processes including cell surface receptor signaling pathway regulation of protein kinase activity and regulation of signal transduction Acts upstream of or within intracellular signal transduction Located in cell projection cytosol and plasma membrane Implicated in prostate cancer Biomarker of lung non-small cell carcinoma
Catalog No. 50-000-13971