Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC phospho-PI3 Kinase p110 beta (Ser1070) Antibody | 10 UL
SDP

Catalog No. 5000442962
Encompass_Preferred

This antibody is a Rabbit-derived and non-conjugated IgG polyclonal antibody, specifically targeting phospho-PI3 Kinase p110 beta (Ser1070).

  • Host: Rabbit
  • Clonality: Polyclonal
  • Species reactivity: Human, mouse, rat
  • Application: WB (1:500-2000), IHC-P (1:100-500), IHC-F (1:100-500), ICC/IF (1:100-500)
  • Purity: Affinity purified
  • Modification: Phosphorylated
  • Isotype: IgG
  • Appearance: Liquid
  • Formulation: Supplied in 0.01M TBS(pH7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
  • Storage: Stored at -20°C for 1 year
  • Avoid repeated freeze / thaw cycles

Catalog No. 50-004-42962 Supplier Medchemexpress LLC Supplier No. HYP8121110UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This antibody is a Rabbit-derived and non-conjugated IgG polyclonal antibody, specifically targeting phospho-PI3 Kinase p110 beta (Ser1070).

  • Host: Rabbit
  • Clonality: Polyclonal
  • Species reactivity: Human, mouse, rat
  • Application: WB (1:500-2000), IHC-P (1:100-500), IHC-F (1:100-500), ICC/IF (1:100-500)
  • Purity: Affinity purified
  • Modification: Phosphorylated
  • Isotype: IgG
  • Appearance: Liquid
  • Formulation: Supplied in 0.01M TBS(pH7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
  • Storage: Stored at -20°C for 1 year
  • Avoid repeated freeze / thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.