Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-PI3 Kinase p85/p55 (Tyr467/Tyr199) Antibody | 100 UL
SDP

Catalog No. 5000442501
Encompass_Preferred

Phospho-PI3 Kinase p85/p55 (Tyr467/Tyr199) Antibody is a non-conjugated liquid antibody from a rabbit host, reactive with human, mouse, and rat. It is supplied at 1.0 mg/mL in a buffer containing PBS, glycerol, BSA, and sodium azide. Suitable for WB, IHC-F, IHC-P, ICC/IF, and ELISA, this antibody should be stored at -20°C for 1 year and shipped with blue ice. For research use only.

  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Concentration: 1.0 mg/mL
  • Conjugation: Non-conjugated
  • Applications: WB, IHC-F, IHC-P, ICC/IF, ELISA
  • Dilution ratios: WB: 1:500-1:1000; IHC-P: 1:50-1:100; IHC-F: 1:50-1:100; ICC/IF: 1:50-1:200; ELISA: 1:10000
  • Formulation: PBS (pH 7.3), 50% glycerol, 0.5% BSA, 0.02% sodium azide (or similar PBS formulation).
  • Storage: -20°C for 1 year; avoid freeze/thaw.
  • Shipping: Blue ice.
  • Predicted band size: 84 kDa
  • Observed band size: 55,85 kDa

Catalog No. 50-004-42501 Supplier Medchemexpress LLC Supplier No. HYP80846100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phospho-PI3 Kinase p85/p55 (Tyr467/Tyr199) Antibody is a non-conjugated liquid antibody from a rabbit host, reactive with human, mouse, and rat. It is supplied at 1.0 mg/mL in a buffer containing PBS, glycerol, BSA, and sodium azide. Suitable for WB, IHC-F, IHC-P, ICC/IF, and ELISA, this antibody should be stored at -20°C for 1 year and shipped with blue ice. For research use only.

  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Concentration: 1.0 mg/mL
  • Conjugation: Non-conjugated
  • Applications: WB, IHC-F, IHC-P, ICC/IF, ELISA
  • Dilution ratios: WB: 1:500-1:1000; IHC-P: 1:50-1:100; IHC-F: 1:50-1:100; ICC/IF: 1:50-1:200; ELISA: 1:10000
  • Formulation: PBS (pH 7.3), 50% glycerol, 0.5% BSA, 0.02% sodium azide (or similar PBS formulation).
  • Storage: -20°C for 1 year; avoid freeze/thaw.
  • Shipping: Blue ice.
  • Predicted band size: 84 kDa
  • Observed band size: 55,85 kDa

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.