Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-PKC zeta (Thr560) Antibody (YA162) | 100 UL
SDP

Catalog No. 5000397157
Encompass_Preferred

Phospho-PKC zeta (Thr560) Antibody (YA162) is a rabbit-derived, non-conjugated IgG monoclonal antibody that targets Phospho-PKC zeta (Thr560).

  • Host: Rabbit
  • Reactivity: Human, mouse
  • Applications: WB, IHC-P
  • Conjugation: Non-conjugated
  • Clonality: Monoclonal antibody
  • Immunogen: Phospho-PKC zeta (Thr560)
  • Isotype: IgG

Catalog No. 50-003-97157 Supplier Medchemexpress LLC Supplier No. HYP80280100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phospho-PKC zeta (Thr560) Antibody (YA162) is a rabbit-derived, non-conjugated IgG monoclonal antibody that targets Phospho-PKC zeta (Thr560).

  • Host: Rabbit
  • Reactivity: Human, mouse
  • Applications: WB, IHC-P
  • Conjugation: Non-conjugated
  • Clonality: Monoclonal antibody
  • Immunogen: Phospho-PKC zeta (Thr560)
  • Isotype: IgG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.