Learn More
Cell Signaling Technology Phospho-PLK1 (Thr210) Matched Antibody Pair, Human, Unconjugated, 1 Kit
SDP
Catalog No. 503132794
Encompass
The Phospho-PLK1 (Thr210) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with custom labeling, such as fluorophores, lanthanides, biotin, and beads. This kit features capture and detection antibodies that bind to non-overlapping epitopes and are supplied in a BSA and azide-free formulation.
  • Compatible with platforms including MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex.
  • Consists of Phospho-PLK1 (Thr210) (D5H7) and PLK1 (208G4) rabbit monoclonal antibodies.
  • Optimized for human species reactivity.
  • Supplied in 1X PBS, pH 7.8.
  • For research use only.
Catalog No. 50-313-2794 Supplier Cell Signaling Technology Supplier No. 45199S
The Phospho-PLK1 (Thr210) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with custom labeling, such as fluorophores, lanthanides, biotin, and beads. This kit features capture and detection antibodies that bind to non-overlapping epitopes and are supplied in a BSA and azide-free formulation.
  • Compatible with platforms including MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex.
  • Consists of Phospho-PLK1 (Thr210) (D5H7) and PLK1 (208G4) rabbit monoclonal antibodies.
  • Optimized for human species reactivity.
  • Supplied in 1X PBS, pH 7.8.
  • For research use only.