Learn More
Cell Signaling Technology Phospho-Rb (Ser780) Matched Antibody Pair, Human, Unconjugated, 1 Kit
SDP
Catalog No. 503131537
Encompass
The Phospho-Rb (Ser780) Matched Antibody Pair is designed for immunoassay technologies and high-throughput ELISA platforms requiring antibody pairs with custom labeling capabilities. This kit provides the necessary capture and detection antibodies for targeted detection of phosphorylated retinoblastoma protein at Ser780.
  • Ideal for use with platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Compatible with diverse labeling methods such as fluorophores, lanthanides, biotin, and beads.
  • Supplied in BSA and azide-free formulation for optimal labeling efficiency.
  • Includes capture and detection antibodies that bind to non-overlapping epitopes.
  • Validated for human species reactivity.
  • Store at -20°C.
Catalog No. 50-313-1537 Supplier Cell Signaling Technology Supplier No. 84096S
The Phospho-Rb (Ser780) Matched Antibody Pair is designed for immunoassay technologies and high-throughput ELISA platforms requiring antibody pairs with custom labeling capabilities. This kit provides the necessary capture and detection antibodies for targeted detection of phosphorylated retinoblastoma protein at Ser780.
  • Ideal for use with platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Compatible with diverse labeling methods such as fluorophores, lanthanides, biotin, and beads.
  • Supplied in BSA and azide-free formulation for optimal labeling efficiency.
  • Includes capture and detection antibodies that bind to non-overlapping epitopes.
  • Validated for human species reactivity.
  • Store at -20°C.